missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenoxy)isobutyric Acid 97.0+%, TCI America™

Catalog No. C094025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C094025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C094025G Supplier TCI America Supplier No. C094025G
Only null left

Chemical Identifiers

CAS 882-09-7
Molecular Formula C10H11ClO3
Molecular Weight (g/mol) 214.645
MDL Number MFCD00004192
InChI Key TXCGAZHTZHNUAI-UHFFFAOYSA-N
Synonym clofibric acid, 2-4-chlorophenoxy-2-methylpropanoic acid, chlorofibrinic acid, clofibrinic acid, chlorfibrinic acid, clofibrin, chlorophibrinic acid, pcib, 2-4-chlorophenoxy-2-methylpropionic acid, clofibrate free acid
PubChem CID 2797
ChEBI CHEBI:34648
IUPAC Name 2-(4-chlorophenoxy)-2-methylpropanoic acid
SMILES CC(C)(C(=O)O)OC1=CC=C(C=C1)Cl

Specifications

Melting Point 120°C
Quantity 25 g
Formula Weight 214.65
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Chlorophenoxy)isobutyric Acid
RTECSNumber UE9455000
TSCA No