missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorobutoxy)tetrahydropyran 95.0+%, TCI America™

Catalog No. p-7124890
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
25 g
5 g
Catalog No. Quantity
C12791G 1 g
C12795G 5 g
C127925G 25 g
3 options

Catalog No. C12791G

Supplier: TCI America C12791G

Only null left

Chemical Identifiers

CAS 41302-05-0
Molecular Formula C9H17ClO2
Molecular Weight (g/mol) 192.68
MDL Number MFCD00142812
InChI Key SZZINZURZKQZLG-UHFFFAOYNA-N
PubChem CID 170488
IUPAC Name 2-(4-chlorobutoxy)oxane
SMILES ClCCCCOC1CCCCO1

Specifications

Color Yellow
Boiling Point 131°C
Quantity 1 g
Formula Weight 192.68
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(4-Chlorobutoxy)tetrahydropyran