missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorobutoxy)tetrahydropyran 95.0+%, TCI America™

Catalog No. C12791G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12791G 1 g
C12795G 5 g
C127925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C12791G Supplier TCI America Supplier No. C12791G
Only null left

Chemical Identifiers

CAS 41302-05-0
Molecular Formula C9H17ClO2
Molecular Weight (g/mol) 192.68
MDL Number MFCD00142812
InChI Key SZZINZURZKQZLG-UHFFFAOYNA-N
PubChem CID 170488
IUPAC Name 2-(4-chlorobutoxy)oxane
SMILES ClCCCCOC1CCCCO1

Specifications

Color Yellow
Boiling Point 131°C
Quantity 1 g
Formula Weight 192.68
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(4-Chlorobutoxy)tetrahydropyran