missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chloro-3-nitrobenzoyl)benzoic Acid 97.0+%, TCI America™

Catalog No. C101325G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C101325G 25 g
C1013250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C101325G Supplier TCI America Supplier No. C101325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 85-54-1
Molecular Formula C14H8ClNO5
Molecular Weight (g/mol) 305.67
MDL Number MFCD00007082
InChI Key RITAQDHCJBLSSL-UHFFFAOYSA-N
Synonym 2-4-chloro-3-nitrobenzoyl benzoic acid, benzoic acid, 2-4-chloro-3-nitrobenzoyl, 3'-nitro-4'-chlorobenzoylbenzoic acid, benzoic acid, o-4-chloro-3-nitrobenzoyl, 2-4-chloro-3-nitrophenyl carbonyl benzoic acid, acmc-209q6m, ksc496o4t, o-4-chloro-3-nitrobenzoyl benzoic acid, 2-4-chloro-3-nitro-benzoyl benzoic acid, 2-4'-chloro-3'-nitrobenzoyl benzoic acid
PubChem CID 66562
IUPAC Name 2-(4-chloro-3-nitrobenzoyl)benzoic acid
SMILES OC(=O)C1=CC=CC=C1C(=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O

Specifications

Melting Point 200°C
Color White-Yellow
Quantity 25 g
Formula Weight 305.67
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Chloro-3-nitrobenzoyl)benzoic Acid