missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chloro-3-nitrobenzoyl)benzoic acid, 98%

Catalog No. AAA1360518
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1360518 50 g
AAA1360530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1360518 Supplier Thermo Scientific Chemicals Supplier No. A1360518
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 85-54-1 | C14H8ClNO5 | 305.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 85-54-1
Molecular Formula C14H8ClNO5
Molecular Weight (g/mol) 305.67
MDL Number MFCD00007082
InChI Key RITAQDHCJBLSSL-UHFFFAOYSA-N
Synonym 2-4-chloro-3-nitrobenzoyl benzoic acid, benzoic acid, 2-4-chloro-3-nitrobenzoyl, 3'-nitro-4'-chlorobenzoylbenzoic acid, benzoic acid, o-4-chloro-3-nitrobenzoyl, 2-4-chloro-3-nitrophenyl carbonyl benzoic acid, acmc-209q6m, ksc496o4t, o-4-chloro-3-nitrobenzoyl benzoic acid, 2-4-chloro-3-nitro-benzoyl benzoic acid, 2-4'-chloro-3'-nitrobenzoyl benzoic acid
PubChem CID 66562
IUPAC Name 2-(4-chloro-3-nitrobenzoyl)benzoic acid
SMILES OC(=O)C1=CC=CC=C1C(=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O

Specifications

Melting Point 198°C to 201°C
Quantity 50 g
Formula Weight 305.68
Percent Purity 98%
Chemical Name or Material 2-(4-Chloro-3-nitrobenzoyl)benzoic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 201-613-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only