missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenyl)naphthalene 98.0+%, TCI America™

Catalog No. B41591G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B41591G Supplier TCI America Supplier No. B41591G
Only null left

Chemical Identifiers

CAS 22082-99-1
Molecular Formula C16H11Br
Molecular Weight (g/mol) 283.17
MDL Number MFCD09271778
InChI Key SAODOTSIOILVSO-UHFFFAOYSA-N
PubChem CID 5461752
IUPAC Name 2-(4-bromophenyl)naphthalene
SMILES BrC1=CC=C(C=C1)C1=CC2=CC=CC=C2C=C1

Specifications

Melting Point 133°C
Quantity 1 g
Formula Weight 283.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Bromophenyl)naphthalene