missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenyl)imidazo[1,2-a]pyridine 98.0+%, TCI America™

Catalog No. B48711G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48711G 1 g
B48715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48711G Supplier TCI America Supplier No. B48711G
Only null left

Chemical Identifiers

CAS 34658-66-7
Molecular Formula C13H9BrN2
Molecular Weight (g/mol) 273.13
MDL Number MFCD00218250
InChI Key GRZUOGFRIHABDK-UHFFFAOYSA-N
Synonym 2-4-bromophenyl imidazo 1,2-a pyridine, 2-4-bromo-phenyl-imidazo 1,2-a pyridine, 2-4-bromophenyl h-imidazo 1,2-a pyridine, acmc-1aihf, 2-4-bromophenyl-imidazo 1,2-a pyridine, 2-4-bromophenyl imidazo 1,2-? pyridine, 2-4-bromophenyl imidazo 1,2-a pyridine #, imidazo 1,2-a pyridine,2-4-bromophenyl, imidazol 1,2-a pyridine, 2-4-bromophenyl
PubChem CID 623416
IUPAC Name 2-(4-bromophenyl)imidazo[1,2-a]pyridine
SMILES BrC1=CC=C(C=C1)C1=CN2C=CC=CC2=N1

Specifications

Melting Point 217°C
Quantity 1 g
Formula Weight 273.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Bromophenyl)imidazo[1,2-a]pyridine