CAS RN 34658-66-7
IUPAC Name:
2-(4-bromophenyl)imidazo[1,2-a]pyridine
Synonyms:
2-(4-bromophenyl)imidazo[1,2-a]pyridine
Molecular Weight (g/mol):
273.13
Molecular Formula:
C13H9BrN2
InChi Key:
GRZUOGFRIHABDK-UHFFFAOYSA-N
SMILES:
BrC1=CC=C(C=C1)C1=CN2C=CC=CC2=N1
1
–
1
of
1
results
1
2-(4-Bromophenyl)imidazo[1,2-a]pyridine 98.0+%, TCI America™
CAS: 34658-66-7 Molecular Formula: C13H9BrN2 Molecular Weight (g/mol): 273.13 MDL Number: MFCD00218250 InChI Key: GRZUOGFRIHABDK-UHFFFAOYSA-N Synonym: 2-4-bromophenyl imidazo 1,2-a pyridine,2-4-bromo-phenyl-imidazo 1,2-a pyridine,2-4-bromophenyl h-imidazo 1,2-a pyridine,acmc-1aihf,2-4-bromophenyl-imidazo 1,2-a pyridine,2-4-bromophenyl imidazo 1,2-? pyridine,2-4-bromophenyl imidazo 1,2-a pyridine #,imidazo 1,2-a pyridine,2-4-bromophenyl,imidazol 1,2-a pyridine, 2-4-bromophenyl PubChem CID: 623416 IUPAC Name: 2-(4-bromophenyl)imidazo[1,2-a]pyridine SMILES: BrC1=CC=C(C=C1)C1=CN2C=CC=CC2=N1