missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenyl)ethanol, 98%

Catalog No. p-7021701
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1414303 1 g
AAA1414306 5 g
AAA1414314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1414303 Supplier Thermo Scientific Chemicals Supplier No. A1414303

CAS: 4654-39-1 | C8H9BrO | 201.063 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4654-39-1
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.063
MDL Number MFCD00002897
InChI Key PMOSJSPFNDUAFY-UHFFFAOYSA-N
Synonym 2-4-bromophenyl ethanol, 4-bromophenethyl alcohol, p-bromophenethyl alcohol, benzeneethanol, 4-bromo, 2-4-bromophenyl ethan-1-ol, 2-4-bromo-phenyl-ethanol, 2-4-bromophenyl ethyl alcohol, 2-p-bromophenyl ethanol, phenethyl alcohol, p-bromo, 4-bromobenzene ethanol
PubChem CID 72851
IUPAC Name 2-(4-bromophenyl)ethanol
SMILES C1=CC(=CC=C1CCO)Br

Specifications

Density 1.436
Boiling Point 106°C to 108°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.573
Quantity 1 g
Beilstein 2079929
Formula Weight 201.06
Percent Purity 98%
Chemical Name or Material 2-(4-Bromophenyl)ethanol
Hazard Category H302+H312-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 225-093-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only