missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenyl)-5-phenylthiophene 98.0+%, TCI America™

Catalog No. B5491200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5491200MG 200 mg
B54911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5491200MG Supplier TCI America Supplier No. B5491200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 118621-30-0
Molecular Formula C16H11BrS
Molecular Weight (g/mol) 315.228
MDL Number MFCD21648452
InChI Key YMDCOEOZWHXGTM-UHFFFAOYSA-N
PubChem CID 10313583
IUPAC Name 2-(4-bromophenyl)-5-phenylthiophene
SMILES C1=CC=C(C=C1)C2=CC=C(S2)C3=CC=C(C=C3)Br

Specifications

Melting Point 190°C
Quantity 200 mg
Formula Weight 315.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Bromophenyl)-5-phenylthiophene