missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenyl)-2-propanol 98.0+%, TCI America™

Catalog No. B52741G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52741G 1 g
B52745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B52741G Supplier TCI America Supplier No. B52741G

Chemical Identifiers

CAS 2077-19-2
Molecular Formula C9H11BrO
Molecular Weight (g/mol) 215.09
MDL Number MFCD11870096
InChI Key AOGYBHJTXLXRSM-UHFFFAOYSA-N
Synonym 2-4-bromophenyl propan-2-ol, 2-4-bromophenyl-2-propanol, acmc-209fdx, 2-4-bromophenyl propan-2-o1, 2-4-bromophenyl-propan-2-ol, 2-4-bromo-phenyl propan-2-ol, 2-4-bromo-phenyl-propan-2-ol, p-bromo-alpha,alpha-dimethylbenzyl alcohol, 4-bromo-alpha,alpha-dimethylbenzyl alcohol
PubChem CID 12681920
IUPAC Name 2-(4-bromophenyl)propan-2-ol
SMILES CC(C)(O)C1=CC=C(Br)C=C1

Specifications

Melting Point 46°C
Color White-Yellow
Boiling Point 85°C
Quantity 1 g
Formula Weight 215.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-(4-Bromophenyl)-2-propanol