missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenyl)-2-propanol, 97%

Catalog No. p-7056153
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64919MD 250 mg
AAH6491903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH64919MD Supplier Thermo Scientific Chemicals Supplier No. H64919MD
Only null left

CAS: 2077-19-2 | C9H11BrO | 215.09 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is a medicine intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is a medicine intermediate.

Solubility
Slightly soluble in water (1.4 g/L) (25°C)

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2077-19-2
Molecular Formula C9H11BrO
Molecular Weight (g/mol) 215.09
MDL Number MFCD11870096
InChI Key AOGYBHJTXLXRSM-UHFFFAOYSA-N
Synonym 2-4-bromophenyl propan-2-ol, 2-4-bromophenyl-2-propanol, acmc-209fdx, 2-4-bromophenyl propan-2-o1, 2-4-bromophenyl-propan-2-ol, 2-4-bromo-phenyl propan-2-ol, 2-4-bromo-phenyl-propan-2-ol, p-bromo-alpha,alpha-dimethylbenzyl alcohol, 4-bromo-alpha,alpha-dimethylbenzyl alcohol
PubChem CID 12681920
IUPAC Name 2-(4-bromophenyl)propan-2-ol
SMILES CC(C)(O)C1=CC=C(Br)C=C1

Specifications

Melting Point 40°C to 45°C
Color White
Density 1.356 g/mL
Boiling Point 75°C to 85°C (0.1 mmHg)
Refractive Index 1.552
Quantity 250 mg
Beilstein 1935066
Solubility Information Slightly soluble in water (1.4g/L) (25°C)
Formula Weight 215.09
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-(4-Bromophenyl)-2-propanol
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 816-112-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only