missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Bromophenyl)-1,1-diphenylethylene 98.0+%, TCI America™
$328.83 - $328.83
Chemical Identifiers
| CAS | 18648-66-3 |
|---|---|
| Molecular Formula | C20H15Br |
| Molecular Weight (g/mol) | 335.24 |
| MDL Number | MFCD01790865 |
| InChI Key | HUCFDUOIMSRYAA-UHFFFAOYSA-N |
| PubChem CID | 630600 |
| IUPAC Name | 1-bromo-4-(2,2-diphenylethenyl)benzene |
| SMILES | BrC1=CC=C(C=C(C2=CC=CC=C2)C2=CC=CC=C2)C=C1 |
Chemical Identifiers
| 18648-66-3 | |
| 335.24 | |
| HUCFDUOIMSRYAA-UHFFFAOYSA-N | |
| 1-bromo-4-(2,2-diphenylethenyl)benzene |
| C20H15Br | |
| MFCD01790865 | |
| 630600 | |
| BrC1=CC=C(C=C(C2=CC=CC=C2)C2=CC=CC=C2)C=C1 |
Specifications
| 18648-66-3 | |
| White-Yellow | |
| MFCD01790865 | |
| HUCFDUOIMSRYAA-UHFFFAOYSA-N | |
| 1-bromo-4-(2,2-diphenylethenyl)benzene | |
| 630600 | |
| ≥98.0% (GC) | |
| 2-(4-Bromophenyl)-1,1-diphenylethylene |
| 79°C | |
| C20H15Br | |
| 5 g | |
| BrC1=CC=C(C=C(C2=CC=CC=C2)C2=CC=CC=C2)C=C1 | |
| 335.24 | |
| 335.24 | |
| Crystalline Powder |
Safety and Handling
TSCA : No