missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenoxy)tetrahydropyran, 98%

Catalog No. p-7028538
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2591409 10 g
AAH2591418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2591409 Supplier Thermo Scientific Chemicals Supplier No. H2591409

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 36603-49-3 | C11H13BrO2 | 257.127 g/mol

2-(4-Bromophenoxy)tetrahydropyran can be used to produce 4-bromo-phenol at temperature of 20°C. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(4-Bromophenoxy)tetrahydropyran can be used to produce 4-bromo-phenol at temperature of 20°C. It is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.

Chemical Identifiers

CAS 36603-49-3
Molecular Formula C11H13BrO2
Molecular Weight (g/mol) 257.127
MDL Number MFCD00091551
InChI Key MXDQGXMBJCGRCB-UHFFFAOYSA-N
Synonym 2-4-bromophenoxy tetrahydro-2h-pyran, 2-4-bromophenoxy tetrahydropyran, 2-4-bromophenoxy oxane, 2h-pyran, 2-4-bromophenoxy tetrahydro, 2-4'-bromophenoxy-tetrahydropyran, 4-bromophenyl thp ether, acmc-209inn, bromophenoxy tetrahydro-2h-pyran, 2-4-bromophenoxy-tetrahydropyran
PubChem CID 4646436
IUPAC Name 2-(4-bromophenoxy)oxane
SMILES C1CCOC(C1)OC2=CC=C(C=C2)Br

Specifications

Melting Point 55°C to 58°C
Quantity 10 g
Solubility Information Slightly soluble in water.
Formula Weight 257.13
Percent Purity 98%
Chemical Name or Material 2-(4-Bromophenoxy)tetrahydropyran
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only