missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Bis(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$85.75 - $85.75
Specifications
| CAS | 153254-09-2 |
|---|---|
| Color | White-Yellow |
| Molecular Formula | C8H5BF6O2 |
| MDL Number | MFCD01631349 |
| InChI Key | WLYPBMBWKYALCG-UHFFFAOYSA-N |
Specifications
| 153254-09-2 | |
| C8H5BF6O2 | |
| WLYPBMBWKYALCG-UHFFFAOYSA-N | |
| 2782667 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD01631349 | |
| [2,4-bis(trifluoromethyl)phenyl]boronic acid | |
| 257.93 |