missing translation for 'onlineSavingsMsg'
Learn More

2,4-Bis(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. B30221G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B30221G Supplier TCI America Supplier No. B30221G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 2,4-Bis(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 153254-09-2
Color White-Yellow
Molecular Formula C8H5BF6O2
Quantity 1 g
MDL Number MFCD01631349
Synonym 2,4-bis trifluoromethyl phenylboronic acid, 2,4-bis trifluoromethyl benzeneboronic acid, 2,4-bis trifluoromethyl phenylboronicacid, 2,4-bis trifluoromethyl phenyl boronic acid, boronic acid, 2,4-bis trifluoromethyl phenyl, 2,4-bis-trifluoromethylphenylboronic acid, pubchem5262, acmc-209d8w, 2,4-bis trifluoromethyl phenyl, 2,4-bistrifluoromethylbenzeneboronic acid
InChI Key WLYPBMBWKYALCG-UHFFFAOYSA-N
SMILES OB(O)C1=CC=C(C=C1C(F)(F)F)C(F)(F)F
IUPAC Name [2,4-bis(trifluoromethyl)phenyl]boronic acid
Molecular Weight (g/mol) 257.93
PubChem CID 2782667
Formula Weight 257.93
Physical Form Crystalline Powder
Show More Show Less