missing translation for 'onlineSavingsMsg'
Learn More

2,4-Bis(trifluoromethyl)benzeneboronic acid, 97%

Catalog No. AAL1816106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1816106 5 g
AAL1816103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1816106 Supplier Thermo Scientific Chemicals Supplier No. L1816106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 153254-09-2 | C8H5BF6O2 | 257.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 153254-09-2
Molecular Formula C8H5BF6O2
Molecular Weight (g/mol) 257.93
MDL Number MFCD01631349
InChI Key WLYPBMBWKYALCG-UHFFFAOYSA-N
Synonym 2,4-bis trifluoromethyl phenylboronic acid, 2,4-bis trifluoromethyl benzeneboronic acid, 2,4-bis trifluoromethyl phenylboronicacid, 2,4-bis trifluoromethyl phenyl boronic acid, boronic acid, 2,4-bis trifluoromethyl phenyl, 2,4-bis-trifluoromethylphenylboronic acid, pubchem5262, acmc-209d8w, 2,4-bis trifluoromethyl phenyl, 2,4-bistrifluoromethylbenzeneboronic acid
PubChem CID 2782667
IUPAC Name [2,4-bis(trifluoromethyl)phenyl]boronic acid
SMILES OB(O)C1=CC=C(C=C1C(F)(F)F)C(F)(F)F

Specifications

Quantity 5 g
Formula Weight 257.93
Percent Purity 97%
Chemical Name or Material 2,4-Bis(trifluoromethyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only