missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Bis(alpha,alpha-dimethylbenzyl)phenol 97.0+%, TCI America™
$33.56 - $165.90
Chemical Identifiers
| CAS | 2772-45-4 |
|---|---|
| Molecular Formula | C24H26O |
| Molecular Weight (g/mol) | 330.471 |
| InChI Key | FMUYQRFTLHAARI-UHFFFAOYSA-N |
| Synonym | 2,4-Di-alpha-cumylphenol |
| PubChem CID | 76013 |
| IUPAC Name | 2,4-bis(2-phenylpropan-2-yl)phenol |
| SMILES | CC(C)(C1=CC=CC=C1)C2=CC(=C(C=C2)O)C(C)(C)C3=CC=CC=C3 |
Chemical Identifiers
| 2772-45-4 | |
| 330.471 | |
| 2,4-Di-alpha-cumylphenol | |
| 2,4-bis(2-phenylpropan-2-yl)phenol |
| C24H26O | |
| FMUYQRFTLHAARI-UHFFFAOYSA-N | |
| 76013 | |
| CC(C)(C1=CC=CC=C1)C2=CC(=C(C=C2)O)C(C)(C)C3=CC=CC=C3 |
Specifications
| 2772-45-4 | |
| White-Yellow | |
| C24H26O | |
| 2,4-Di-alpha-cumylphenol | |
| CC(C)(C1=CC=CC=C1)C2=CC(=C(C=C2)O)C(C)(C)C3=CC=CC=C3 | |
| 330.471 | |
| 330.47 | |
| Crystalline Powder |
| 65°C | |
| 221°C | |
| 25 g | |
| FMUYQRFTLHAARI-UHFFFAOYSA-N | |
| 2,4-bis(2-phenylpropan-2-yl)phenol | |
| 76013 | |
| ≥97.0% (GC) | |
| 2,4-Bis(alpha,alpha-dimethylbenzyl)phenol |
Safety and Handling
TSCA : Yes