missing translation for 'onlineSavingsMsg'
Learn More

2,4'-Bipyridyl 97.0+%, TCI America™

Catalog No. B08631G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B08631G 1 g
B08635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B08631G Supplier TCI America Supplier No. B08631G

Chemical Identifiers

CAS 581-47-5
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.188
MDL Number MFCD00006217
InChI Key RMHQDKYZXJVCME-UHFFFAOYSA-N
Synonym 2,4'-bipyridine, 2,4'-dipyridyl, 4-pyridylpyridine, 2,4'-bipyridyl, 2,4-bipyridyl, 2,4'-dipyridine, unii-c6sn6ru851, ccris 3428, 2-4-pyridyl pyridine, 4-pyridyl-2-pyridine
PubChem CID 68488
IUPAC Name 2-pyridin-4-ylpyridine
SMILES C1=CC=NC(=C1)C2=CC=NC=C2

Specifications

Melting Point 64°C
Boiling Point 110°C
Quantity 1 g
Formula Weight 156.19
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4′-Bipyridyl