accessibility menu, dialog, popup

UserName

NN

CAS RN 581-47-5

IUPAC Name: 2,4'-bipyridine
Synonyms: (4-pyridyl)-2-pyridine
Molecular Weight (g/mol): 156.19
Molecular Formula: C10H8N2
InChi Key: RMHQDKYZXJVCME-UHFFFAOYSA-N
SMILES: C1=CC=C(N=C1)C1=CC=NC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)