missing translation for 'onlineSavingsMsg'
Learn More

2,4'-Bipyridine, 97%

Catalog No. p-7029854
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27366MD 250 mg
AAH2736603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH27366MD Supplier Thermo Scientific Chemicals Supplier No. H27366MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 581-47-5 | C10H8N2 | 156.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 581-47-5
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.188
MDL Number MFCD00006217
InChI Key RMHQDKYZXJVCME-UHFFFAOYSA-N
Synonym 2,4'-bipyridine, 2,4'-dipyridyl, 4-pyridylpyridine, 2,4'-bipyridyl, 2,4-bipyridyl, 2,4'-dipyridine, unii-c6sn6ru851, ccris 3428, 2-4-pyridyl pyridine, 4-pyridyl-2-pyridine
PubChem CID 68488
IUPAC Name 2-pyridin-4-ylpyridine
SMILES C1=CC=NC(=C1)C2=CC=NC=C2

Specifications

Melting Point 59°C to 63°C
Boiling Point 281°C
Quantity 250 mg
UN Number UN2811
Formula Weight 156.19
Percent Purity 97%
Chemical Name or Material 2,4'-Bipyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 209-467-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only