missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Biphenylylcarbonyl)benzoic Acid 97.0+%, TCI America™

$77.48 - $185.19
Chemical Identifiers
CAS | 42797-18-2 |
---|---|
Molecular Formula | C20H14O3 |
Molecular Weight (g/mol) | 302.33 |
MDL Number | MFCD00392368 |
InChI Key | NBYNVBRXIUUSIG-UHFFFAOYSA-N |
PubChem CID | 39302 |
IUPAC Name | 2-{[1,1'-biphenyl]-4-carbonyl}benzoic acid |
SMILES | OC(=O)C1=CC=CC=C1C(=O)C1=CC=C(C=C1)C1=CC=CC=C1 |
Chemical Identifiers
42797-18-2 | |
302.33 | |
NBYNVBRXIUUSIG-UHFFFAOYSA-N | |
2-{[1,1'-biphenyl]-4-carbonyl}benzoic acid |
C20H14O3 | |
MFCD00392368 | |
39302 | |
OC(=O)C1=CC=CC=C1C(=O)C1=CC=C(C=C1)C1=CC=CC=C1 |
Specifications
42797-18-2 | |
White | |
MFCD00392368 | |
NBYNVBRXIUUSIG-UHFFFAOYSA-N | |
2-{[1,1'-biphenyl]-4-carbonyl}benzoic acid | |
39302 | |
≥97.0% (GC,T) | |
2-(4-Biphenylylcarbonyl)benzoic Acid |
229°C | |
C20H14O3 | |
1 g | |
OC(=O)C1=CC=CC=C1C(=O)C1=CC=C(C=C1)C1=CC=CC=C1 | |
302.33 | |
302.33 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : DG4375000
TSCA : Yes