missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Aminophenyl)ethanol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 108°C |
| Boiling Point | 255°C |
| Quantity | 25 g |
| Formula Weight | 137.18 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-(4-Aminophenyl)ethanol |
Safety and Handling
| EINECSNumber | (3)-1071 |
| TSCA | Yes |