accessibility menu, dialog, popup

UserName

NH2OH

CAS RN 104-10-9

IUPAC Name: 2-(4-aminophenyl)ethan-1-ol
Synonyms: P-aminophenylethanol
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: QXHDYMUPPXAMPQ-UHFFFAOYSA-N
SMILES: NC1=CC=C(CCO)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (4)
  • (1)
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)