missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Aminophenyl)ethanol, 97%

Catalog No. aal1522306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1522306 5 g
AAL1522314 25 g
AAL1522322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1522306 Supplier Thermo Scientific Chemicals Supplier No. L1522306
Only null left

CAS: 104-10-9 | C8H11NO | 137.182 g/mol

2-(4-Aminophenyl)ethanol is used as nonsymmetric monomer in the preparation of ordered [head-to-head (H-H) or tail-to-tail (T-T)] poly(amide-ester), in the synthesis of 4-aminostyrene and in functionalization of graphene nanoplatelets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(4-Aminophenyl)ethanol is used as nonsymmetric monomer in the preparation of ordered [head-to-head (H-H) or tail-to-tail (T-T)] poly(amide-ester), in the synthesis of 4-aminostyrene and in functionalization of graphene nanoplatelets.

Solubility
Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. It is sensitive to air. Incompatible with strong oxidizing agents. Store under dry inert gas. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 104-10-9
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00007922
InChI Key QXHDYMUPPXAMPQ-UHFFFAOYSA-N
Synonym 2-4-aminophenyl ethanol, 4-aminophenethyl alcohol, p-aminophenethyl alcohol, p-aminophenylethanol, benzeneethanol, 4-amino, 2-p-aminophenyl ethanol, 2-4-aminophenyl ethan-1-ol, 4-2-hydroxyethyl aniline, benzeneethanol, ar-amino, p-2-hydroxyethyl aniline
PubChem CID 66904
IUPAC Name 2-(4-aminophenyl)ethanol
SMILES C1=CC(=CC=C1CCO)N

Specifications

Melting Point 108°C to 110°C
Quantity 5 g
Beilstein 907205
Solubility Information Soluble in methanol.
Formula Weight 137.18
Percent Purity 97%
Chemical Name or Material 2-(4-Aminophenyl)ethanol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 203-174-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only