missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Amino-2-nitroanilino)ethanol 95.0+%, TCI America™

Catalog No. p-7118015
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26715G 5 g
A267125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26715G Supplier TCI America Supplier No. A26715G
Only null left

Chemical Identifiers

CAS 2871-01-4
Molecular Formula C8H11N3O3
Molecular Weight (g/mol) 197.19
MDL Number MFCD00071801
InChI Key GZGZVOLBULPDFD-UHFFFAOYSA-N
Synonym 4-Amino-N-(2-hydroxyethyl)-2-nitroaniline, N1-(2-Hydroxyethyl)-2-nitro-1,4-phenylenediamine
PubChem CID 3465817
ChEBI CHEBI:82473
IUPAC Name 2-(4-amino-2-nitroanilino)ethanol
SMILES C1=CC(=C(C=C1N)[N+](=O)[O-])NCCO

Specifications

Melting Point 126°C
Color Black-Green
Quantity 5 g
Formula Weight 197.19
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Amino-2-nitroanilino)ethanol
RTECSNumber KJ6500000
TSCA Yes