missing translation for 'onlineSavingsMsg'
Learn More

2,4,7-Triamino-6-phenylpteridine, 98%

Catalog No. aab2004414
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2004414 25 g
AAB2004406 5 g
AAB2004422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2004414 Supplier Thermo Scientific Chemicals Supplier No. B2004414
Only null left

CAS: 396-01-0 | C12H11N7 | 253.269 g/mol

Weak diuretic with potassium sparing properties that blocks sodium reuptake in the kidneys

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 396-01-0
Molecular Formula C12H11N7
Molecular Weight (g/mol) 253.269
MDL Number MFCD00006708
InChI Key FNYLWPVRPXGIIP-UHFFFAOYSA-N
Synonym triamterene, 2,4,7-triamino-6-phenylpteridine, dyrenium, triamteren, dytac, pterofen, pterophene, triamteril, triteren, ademin
PubChem CID 5546
IUPAC Name 6-phenylpteridine-2,4,7-triamine
SMILES C1=CC=C(C=C1)C2=NC3=C(N=C2N)N=C(N=C3N)N

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 266723
Merck Index 14,9599
Formula Weight 253.27
Percent Purity 98%
Chemical Name or Material 2,4,7-Triamino-6-phenylpteridine
Hazard Category H302-H315-H319-H335-H351
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
EINECSNumber 206-904-3
RTECSNumber UO3470000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only