accessibility menu, dialog, popup

UserName

CAS RN 396-01-0

IUPAC Name: 6-phenylpteridine-2,4,7-triamine
Synonyms: triamterene tricilone jorba brand of triamterene renezide trispan triamterene preparation reviten apo-triazide triamthiazid sali-puren
Molecular Weight (g/mol): 253.27
Molecular Formula: C12H11N7
InChi Key: FNYLWPVRPXGIIP-UHFFFAOYSA-N
SMILES: NC1=NC(N)=C2N=C(C(N)=NC2=N1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results