Learn More
2,4,7-Triamino-6-phenylpteridine, 98%
CAS: 396-01-0 | C12H11N7 | 253.269 g/mol
Supplier: Thermo Scientific Chemicals B2004406
Description
Weak diuretic with potassium sparing properties that blocks sodium reuptake in the kidneys
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2,4,7-Triamino-6-phenylpteridine | |
| >300°C | |
| MFCD00006708 | |
| 266723 | |
| triamterene, 2,4,7-triamino-6-phenylpteridine, dyrenium, triamteren, dytac, pterofen, pterophene, triamteril, triteren, ademin | |
| C1=CC=C(C=C1)C2=NC3=C(N=C2N)N=C(N=C3N)N | |
| 253.269 | |
| 253.27 |
| 396-01-0 | |
| C12H11N7 | |
| 5 g | |
| 14,9599 | |
| FNYLWPVRPXGIIP-UHFFFAOYSA-N | |
| 6-phenylpteridine-2,4,7-triamine | |
| 5546 | |
| 98% |
Chemical Identifiers
| 396-01-0 | |
| 253.269 | |
| FNYLWPVRPXGIIP-UHFFFAOYSA-N | |
| 5546 | |
| C1=CC=C(C=C1)C2=NC3=C(N=C2N)N=C(N=C3N)N |
| C12H11N7 | |
| MFCD00006708 | |
| triamterene, 2,4,7-triamino-6-phenylpteridine, dyrenium, triamteren, dytac, pterofen, pterophene, triamteril, triteren, ademin | |
| 6-phenylpteridine-2,4,7-triamine |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
H302-H315-H319-H335-H351
EINECSNumber : 206-904-3
RTECSNumber : UO3470000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only