missing translation for 'onlineSavingsMsg'
Learn More

2,4,7,9-Tetramethyl-5-decyne-4,7-diol (DL- and meso- mixture) 95.0+%, TCI America™

Catalog No. T072425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T072425G 25 g
T0724500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T072425G Supplier TCI America Supplier No. T072425G

Chemical Identifiers

CAS 126-86-3
Molecular Formula C14H26O2
Molecular Weight (g/mol) 226.36
MDL Number MFCD00008942
InChI Key LXOFYPKXCSULTL-UHFFFAOYNA-N
Synonym 2,4,7,9-tetramethyl-5-decyne-4,7-diol, surfynol 104, surfynol 104a, surfynol 104e, surfynol tg, syrfynol 104, tetramethyl decynediol, 5-decyne-4,7-diol, 2,4,7,9-tetramethyl, surfynol 104pa, 1,4-diisobutyl-1,4-dimethylbutynediol
PubChem CID 31362
IUPAC Name 2,4,7,9-tetramethyldec-5-yne-4,7-diol
SMILES CC(C)CC(C)(O)C#CC(C)(O)CC(C)C

Specifications

Melting Point 54°C
Color White-Yellow
Boiling Point 260°C
Quantity 25 g
Formula Weight 226.36
Percent Purity ≥95.0% (GC)
Physical Form Wax
Chemical Name or Material 2,4,7,9-Tetramethyl-5-decyne-4,7-diol (DL- and meso- mixture)
EINECSNumber (2)-0263
TSCA Yes