accessibility menu, dialog, popup

UserName

CH3H3CCH3OHCH3OHH3CCH3

CAS RN 126-86-3

IUPAC Name: 2,4,7,9-tetramethyldec-5-yne-4,7-diol
Synonyms: 2,4,7,9-tetramethyldec-5-yne-4,7-diol
Molecular Weight (g/mol): 226.36
Molecular Formula: C14H26O2
InChi Key: LXOFYPKXCSULTL-UHFFFAOYNA-N
SMILES: CC(C)CC(C)(O)C#CC(C)(O)CC(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results