missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trivinylboroxin - Pyridine Complex 98.0+%, TCI America™

Catalog No. p-7137246
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24981G 1 g
T24985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24981G Supplier TCI America Supplier No. T24981G

Chemical Identifiers

CAS 95010-17-6
Molecular Formula C11H14B3NO3
Molecular Weight (g/mol) 240.667
MDL Number MFCD03839440
InChI Key YLHJACXHRQQNQR-UHFFFAOYSA-N
Synonym 2,4,6-trivinyl-1,3,5,2,4,6-trioxatriborinane compound with pyridine 1:1, trivinylboroxin pyridine complex, vinylboronic anhydride pyridine complex, 2,4,6-trivinylcyclotriboroxane-pyridine, 2,4,6-trivinylcyclotriboroxane pyridine, vinylboronic anhydride pyridine, 2,4,6-trivinylcyclotriboroxane pyridine complex, 2,4,6-trivinylcyclotriboroxane-pyridine complex, boroxin, triethenyl-, compd. with pyridine 1:1, pyridine; triethenyl-1,3,5,2,4,6-trioxatriborinane
PubChem CID 2734806
IUPAC Name pyridine;2,4,6-tris(ethenyl)-1,3,5,2,4,6-trioxatriborinane
SMILES B1(OB(OB(O1)C=C)C=C)C=C.C1=CC=NC=C1

Specifications

Melting Point 51°C
Color White-Yellow
Quantity 1 g
Formula Weight 240.67
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Trivinylboroxin - Pyridine Complex