missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tris[bis(methoxymethyl)amino]-1,3,5-triazine 98.0+%, TCI America™

Catalog No. T20595G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20595G 5 g
T205925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T20595G Supplier TCI America Supplier No. T20595G

Chemical Identifiers

CAS 3089-11-0
Molecular Formula C15H30N6O6
Molecular Weight (g/mol) 390.441
MDL Number MFCD00047342
InChI Key BNCADMBVWNPPIZ-UHFFFAOYSA-N
PubChem CID 62479
IUPAC Name 2-N,2-N,4-N,4-N,6-N,6-N-hexakis(methoxymethyl)-1,3,5-triazine-2,4,6-triamine
SMILES COCN(COC)C1=NC(=NC(=N1)N(COC)COC)N(COC)COC

Specifications

Melting Point 52°C
Color White
Quantity 5 g
Synonym N,N,N′C,N′C,N′C′C,N′C′C-Hexakis(methoxymethyl)melamine
Formula Weight 390.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tris[bis(methoxymethyl)amino]-1,3,5-triazine
EINECSNumber (5)-1012&(5)-3839
RTECSNumber XZ1065000
TSCA Yes