accessibility menu, dialog, popup

UserName

CH3ONOCH3NNNNOH3COCH3NOH3COCH3

CAS RN 3089-11-0

IUPAC Name: N2,N2,N4,N4,N6,N6-hexakis(methoxymethyl)-1,3,5-triazine-2,4,6-triamine
Synonyms: N2,N2,N4,N4,N6,N6-hexakis(methoxymethyl)-1,3,5-triazine-2,4,6-triamine
Molecular Weight (g/mol): 390.44
Molecular Formula: C15H30N6O6
InChi Key: BNCADMBVWNPPIZ-UHFFFAOYSA-N
SMILES: COCN(COC)C1=NC(=NC(=N1)N(COC)COC)N(COC)COC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results