missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tris(3,4,5-trifluorophenyl)boroxin 98.0+%, TCI America™

Catalog No. T19295G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19295G 5 g
T19291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T19295G Supplier TCI America Supplier No. T19295G
Only null left

Chemical Identifiers

CAS 223440-94-6
Molecular Formula C18H6B3F9O3
Molecular Weight (g/mol) 473.659
MDL Number MFCD03844809
InChI Key SXINGRFBONUWIF-UHFFFAOYSA-N
Synonym 3,4,5-Trifluorophenylboronic Anhydride, Tris(3,4,5-trifluorophenyl)cycloboroxane
PubChem CID 20654586
IUPAC Name 2,4,6-tris(3,4,5-trifluorophenyl)-1,3,5,2,4,6-trioxatriborinane
SMILES B1(OB(OB(O1)C2=CC(=C(C(=C2)F)F)F)C3=CC(=C(C(=C3)F)F)F)C4=CC(=C(C(=C4)F)F)F

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 473.66
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tris(3,4,5-trifluorophenyl)boroxin