missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Tris(3,4,5-trifluorophenyl)boroxin 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 473.66 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,4,6-Tris(3,4,5-trifluorophenyl)boroxin |
Safety and Handling
| TSCA | No |