missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triphenylnitrobenzene 98.0+%, TCI America™

Catalog No. T051810G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T051810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T051810G Supplier TCI America Supplier No. T051810G
Only null left

Chemical Identifiers

CAS 10368-47-5
Molecular Formula C24H17NO2
Molecular Weight (g/mol) 351.405
MDL Number MFCD00060090
InChI Key VCRVMYZAKLXTFQ-UHFFFAOYSA-N
PubChem CID 3107828
IUPAC Name 2-nitro-1,3,5-triphenylbenzene
SMILES C1=CC=C(C=C1)C2=CC(=C(C(=C2)C3=CC=CC=C3)[N+](=O)[O-])C4=CC=CC=C4

Specifications

Melting Point 147°C
Color White-Yellow
Quantity 10 g
Formula Weight 351.41
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Triphenylnitrobenzene