accessibility menu, dialog, popup

UserName

NOO

CAS RN 10368-47-5

IUPAC Name: 2-nitro-3,5-diphenyl-1,1'-biphenyl
Synonyms: 2-nitro-3,5-diphenyl-1,1'-biphenyl
Molecular Weight (g/mol): 351.41
Molecular Formula: C24H17NO2
InChi Key: VCRVMYZAKLXTFQ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(C=C(C=C1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results