missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylpyrylium Tetrafluoroborate 98.0+%, TCI America™

Catalog No. p-7137184
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23755G 5 g
T237525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23755G Supplier TCI America Supplier No. T23755G
Only null left

Chemical Identifiers

CAS 773-01-3
Molecular Formula C8H11BF4O
Molecular Weight (g/mol) 209.98
MDL Number MFCD00032098
InChI Key PYOPRIFMELCENN-UHFFFAOYSA-N
Synonym 2,4,6-trimethylpyrylium tetrafluoroborate, 2,4,6-trimethyl-1??-pyran-1-ylium tetrafluoroborate, 2,4,6-trimethylpyrylium tetrafluoborate, 2,4,6-trimethylpyryliumtetrafluoroborate, pyrylium, 2,4,6-trimethyl-, tetrafluoroborate 1-, pyrylium, 2,4,6-trimethyl-, tetrafluoroborate 1-1:1, 2,4,6-trimethyl-1$l^ 4-pyran-1-ylium tetrafluoroborate
PubChem CID 164732
IUPAC Name 2,4,6-trimethyl-1λ⁴-pyran-1-ylium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CC1=CC(C)=[O+]C(C)=C1

Specifications

Melting Point 208°C
Color Yellow
Quantity 5 g
UN Number 1759
Formula Weight 209.98
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Trimethylpyrylium Tetrafluoroborate