accessibility menu, dialog, popup

UserName

CH3CH3OH3CFBFFF

CAS RN 773-01-3

IUPAC Name: 2,4,6-trimethyl-1λ⁴-pyran-1-ylium; tetrafluoroboranuide
Synonyms: 2,4,6-trimethyl-1λ⁴-pyran-1-ylium tetrafluoroborate
Molecular Weight (g/mol): 209.98
Molecular Formula: C8H11BF4O
InChi Key: PYOPRIFMELCENN-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CC1=CC(C)=[O+]C(C)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)