missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylpyridine 98.0+%, TCI America™

Catalog No. T071625ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T071625ML 25 mL
T0716100ML 100 mL
T0716500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T071625ML Supplier TCI America Supplier No. T071625ML

Chemical Identifiers

CAS 108-75-8
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00006338
InChI Key BWZVCCNYKMEVEX-UHFFFAOYSA-N
Synonym 2,4,6-collidine, gamma-collidine, s-collidine, pyridine, 2,4,6-trimethyl, collidine, sym-collidine, 2,4,6-kollidin, unii-7ie4bk5j5v, 2,4,6-trimethyl-pyridine, .gamma.-collidine
PubChem CID 7953
IUPAC Name 2,4,6-trimethylpyridine
SMILES CC1=CC(C)=NC(C)=C1

Specifications

Melting Point -43°C
Boiling Point 171°C
Quantity 25 mL
UN Number 1992
Formula Weight 121.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,4,6-Trimethylpyridine
EINECSNumber (5)-0715
RTECSNumber UU0970000
TSCA Yes