missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylphenol 98.0+%, TCI America™

Catalog No. p-7136310
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T048625G 25 g
T0486100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T048625G Supplier TCI America Supplier No. T048625G
Only null left

Chemical Identifiers

CAS 527-60-6
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002235
InChI Key BPRYUXCVCCNUFE-UHFFFAOYSA-N
Synonym mesitol, phenol, 2,4,6-trimethyl, mesityl alcohol, 2-hydroxymesitylene, 2,4,6-trimetylofenol, 1,3,5-trimethylphenol, 2,4,6-trimethyl-phenol, 1-hydroxy-2,4,6-trimethylbenzene, 2,4,6-trimethylofenol, unii-fpz32614n6
PubChem CID 10698
IUPAC Name 2,4,6-trimethylphenol
SMILES CC1=CC(=C(C(=C1)C)O)C

Specifications

Melting Point 72°C
Color White-Yellow
Boiling Point 95°C
Quantity 25 g
UN Number 2430
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Trimethylphenol
EINECSNumber (3)-0540&(4)-0057&(9)-0899&(9)-1783
RTECSNumber OX6590000
TSCA Yes