missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylbenzyl Alcohol 98.0+%, TCI America™

Catalog No. T19055G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19055G 5 g
T190525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T19055G Supplier TCI America Supplier No. T19055G

Chemical Identifiers

CAS 4170-90-5
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00014422
InChI Key LODDFDHPSIYCTK-UHFFFAOYSA-N
Synonym 2,4,6-trimethylbenzyl alcohol, mesitylmethanol, mesitylcarbinol, 2,4,6-trimethylphenyl methanol, unii-rev3kvt84n, benzenemethanol, 2,4,6-trimethyl, 2,4,6-trimethylbenzylic alcohol, benzyl alcohol, 2,4,6-trimethyl, rev3kvt84n, 2,4,6-trimethylphenyl methan-1-ol
PubChem CID 20139
IUPAC Name (2,4,6-trimethylphenyl)methanol
SMILES CC1=CC(=C(C(=C1)C)CO)C

Specifications

Melting Point 88°C
Color White
Boiling Point 141°C
Quantity 5 g
Formula Weight 150.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Trimethylbenzyl Alcohol
RTECSNumber DP1402000
TSCA No