missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylaniline 99.0+%, TCI America™

Catalog No. p-7136298
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
T046725ML 25 mL
T0467100ML 100 mL
T0467500ML 500 mL
3 options

Catalog No. T046725ML

Supplier: TCI America T046725ML

Only null left

Chemical Identifiers

CAS 88-05-1
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00007740
InChI Key KWVPRPSXBZNOHS-UHFFFAOYSA-N
Synonym mesidine, aminomesitylene, mesitylamine, benzenamine, 2,4,6-trimethyl, mesidin, 2,4,6-trimethylbenzenamine, 2-aminomesitylene, 2-amino-1,3,5-trimethylbenzene, 2,4,6-trimethyl aniline, mesitylene, 2-amino
PubChem CID 6913
ChEBI CHEBI:82545
IUPAC Name 2,4,6-trimethylaniline
SMILES CC1=CC(=C(C(=C1)C)N)C

Specifications

Boiling Point 230°C
Quantity 25 mL
UN Number 2810
Formula Weight 135.21
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4,6-Trimethylaniline
EINECSNumber (3)-0132
RTECSNumber BZ0700000
TSCA Yes