accessibility menu, dialog, popup

UserName

ClCH3CH3NH3H3C

CAS RN 88-05-1

IUPAC Name: 2,4,6-trimethylanilinium chloride
Synonyms: 2,4,6-trimethylaniline
Molecular Weight (g/mol): 171.67
Molecular Formula: C9H14ClN
InChi Key: WUYJXWRFOUCHEB-UHFFFAOYSA-N
SMILES: [Cl-].CC1=CC(C)=C([NH3+])C(C)=C1
Boiling Point 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)