missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylaniline 99.0+%, TCI America™

Catalog No. T046725ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T046725ML 25 mL
T0467100ML 100 mL
T0467500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T046725ML Supplier TCI America Supplier No. T046725ML

Chemical Identifiers

CAS 88-05-1
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00007740
InChI Key KWVPRPSXBZNOHS-UHFFFAOYSA-N
Synonym mesidine, aminomesitylene, mesitylamine, benzenamine, 2,4,6-trimethyl, mesidin, 2,4,6-trimethylbenzenamine, 2-aminomesitylene, 2-amino-1,3,5-trimethylbenzene, 2,4,6-trimethyl aniline, mesitylene, 2-amino
PubChem CID 6913
ChEBI CHEBI:82545
IUPAC Name 2,4,6-trimethylaniline
SMILES CC1=CC(=C(C(=C1)C)N)C

Specifications

Boiling Point 230°C
Quantity 25 mL
UN Number 2810
Formula Weight 135.21
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4,6-Trimethylaniline
EINECSNumber (3)-0132
RTECSNumber BZ0700000
TSCA Yes