Learn More
2,4,6-Trimethylaniline, 98%
CAS: 88-05-1 | C9H13N | 135.21 g/mol
Supplier: Thermo Scientific Chemicals A1304936
Description
2,4,6-Trimethylaniline is useful for the preparation of Grubbs' catalyst. It is used as a precursor to dyes. It undergoes condensation reaction with glyoxal to prepare glyoxal-bis(mesitylimine). It is also involved in the preparation of 1,3-diketimines ligands by condensation with 1,3-diketones. Further, it is used for the production of Acid Blue 129 dye, which is useful in histochemistry studies. In addition to this, it serves as a building block to various bulky ligands.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,4,6-Trimethylaniline is useful for the preparation of Grubbs′ catalyst. It is used as a precursor to dyes. It undergoes condensation reaction with glyoxal to prepare glyoxal-bis(mesitylimine). It is also involved in the preparation of 1,3-diketimines ligands by condensation with 1,3-diketones. Further, it is used for the production of Acid Blue 129 dye, which is useful in histochemistry studies. In addition to this, it serves as a building block to various bulky ligands.
Solubility
Slightly miscible with carbon tetrachloride. Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
| 2,4,6-Trimethylaniline | |
| -5°C | |
| 232°C to 234°C | |
| C9H13N | |
| MFCD00007740 | |
| UN2810 | |
| mesidine, aminomesitylene, mesitylamine, benzenamine, 2,4,6-trimethyl, mesidin, 2,4,6-trimethylbenzenamine, 2-aminomesitylene, 2-amino-1,3,5-trimethylbenzene, 2,4,6-trimethyl aniline, mesitylene, 2-amino | |
| KWVPRPSXBZNOHS-UHFFFAOYSA-N | |
| 2,4,6-trimethylaniline | |
| 6913 | |
| 135.21 |
| 88-05-1 | |
| 0.962 | |
| 96°C (204°F) | |
| 1.551 | |
| 500 g | |
| 636559 | |
| Slightly miscible with carbon tetrachloride. Immiscible with water. | |
| CC1=CC(=C(C(=C1)C)N)C | |
| 135.21 | |
| CHEBI:82545 | |
| 98% |
Chemical Identifiers
| 88-05-1 | |
| 135.21 | |
| KWVPRPSXBZNOHS-UHFFFAOYSA-N | |
| 6913 | |
| 2,4,6-trimethylaniline |
| C9H13N | |
| MFCD00007740 | |
| mesidine, aminomesitylene, mesitylamine, benzenamine, 2,4,6-trimethyl, mesidin, 2,4,6-trimethylbenzenamine, 2-aminomesitylene, 2-amino-1,3,5-trimethylbenzene, 2,4,6-trimethyl aniline, mesitylene, 2-amino | |
| CHEBI:82545 | |
| CC1=CC(=C(C(=C1)C)N)C |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P262-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
H301+H311+H331-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 201-794-3
RTECSNumber : BZ0700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only