missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylaniline, 98%

CAS: 88-05-1 | C9H13N | 135.21 g/mol

Supplier:  Thermo Scientific Chemicals A130490E

 View more versions of this product

Catalog No. AAA130490E

Only null left
Explore available promotions

Description

Description

2,4,6-Trimethylaniline is useful for the preparation of Grubbs' catalyst. It is used as a precursor to dyes. It undergoes condensation reaction with glyoxal to prepare glyoxal-bis(mesitylimine). It is also involved in the preparation of 1,3-diketimines ligands by condensation with 1,3-diketones. Further, it is used for the production of Acid Blue 129 dye, which is useful in histochemistry studies. In addition to this, it serves as a building block to various bulky ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethylaniline is useful for the preparation of Grubbs′ catalyst. It is used as a precursor to dyes. It undergoes condensation reaction with glyoxal to prepare glyoxal-bis(mesitylimine). It is also involved in the preparation of 1,3-diketimines ligands by condensation with 1,3-diketones. Further, it is used for the production of Acid Blue 129 dye, which is useful in histochemistry studies. In addition to this, it serves as a building block to various bulky ligands.

Solubility
Slightly miscible with carbon tetrachloride. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

2,4,6-Trimethylaniline
-5°C
232°C to 234°C
C9H13N
MFCD00007740
UN2810
mesidine, aminomesitylene, mesitylamine, benzenamine, 2,4,6-trimethyl, mesidin, 2,4,6-trimethylbenzenamine, 2-aminomesitylene, 2-amino-1,3,5-trimethylbenzene, 2,4,6-trimethyl aniline, mesitylene, 2-amino
KWVPRPSXBZNOHS-UHFFFAOYSA-N
2,4,6-trimethylaniline
6913
135.21
88-05-1
0.962
96°C (204°F)
1.551
2500 g
636559
Slightly miscible with carbon tetrachloride. Immiscible with water.
CC1=CC(=C(C(=C1)C)N)C
135.21
CHEBI:82545
98%

Chemical Identifiers

88-05-1
135.21
KWVPRPSXBZNOHS-UHFFFAOYSA-N
6913
2,4,6-trimethylaniline
C9H13N
MFCD00007740
mesidine, aminomesitylene, mesitylamine, benzenamine, 2,4,6-trimethyl, mesidin, 2,4,6-trimethylbenzenamine, 2-aminomesitylene, 2-amino-1,3,5-trimethylbenzene, 2,4,6-trimethyl aniline, mesitylene, 2-amino
CHEBI:82545
CC1=CC(=C(C(=C1)C)N)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P262-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c

H301+H311+H331-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 201-794-3

RTECSNumber : BZ0700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only