missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylaniline, 97%

Catalog No. ac157960025
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
2.5 kg
500 g
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC157960025 2.5 kg Glass bottle
AC157961000 100 g Glass bottle
AC157965000 500 g Glass bottle
3 options

Catalog No. AC157960025

Supplier: Thermo Scientific Chemicals 157960025

Only null left

CAS: 88-05-1 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88-05-1
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00007740
InChI Key KWVPRPSXBZNOHS-UHFFFAOYSA-N
Synonym mesidine, aminomesitylene, mesitylamine, benzenamine, 2,4,6-trimethyl, mesidin, 2,4,6-trimethylbenzenamine, 2-aminomesitylene, 2-amino-1,3,5-trimethylbenzene, 2,4,6-trimethyl aniline, mesitylene, 2-amino
PubChem CID 6913
ChEBI CHEBI:82545
IUPAC Name 2,4,6-trimethylaniline
SMILES CC1=CC(=C(C(=C1)C)N)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -5.0°C
Density 0.9600g/mL
Boiling Point 233.0°C
Flash Point 96°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5505 to 1.5525
Linear Formula (CH3)3C6H2NH2
Quantity 2.5 kg
Beilstein 12,116
Specific Gravity 0.96
Solubility Information Solubility in water: immiscible with water. Other solubilities: slightly soluble in carbon tetrachloride,soluble in most common solvents
Formula Weight 135.21
Percent Purity 97%
Chemical Name or Material 2, 4, 6-Trimethylaniline
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-794-3

RUO – Research Use Only