missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trimethyl-1,3-phenylenediamine 98.0+%, TCI America™
Supplier: TCI America T1275100G
Specifications
| 2,4,6-Trimethyl-1,3-phenylenediamine | |
| 91°C | |
| MFCD00010149 | |
| 2,4-Diaminomesitylene, 2,4-Diamino-1,3,5-trimethylbenzene | |
| CC1=CC(C)=C(N)C(C)=C1N | |
| 150.23 | |
| 150.23 | |
| Crystalline Powder |
| 3102-70-3 | |
| C9H14N2 | |
| 100 g | |
| ZVDSMYGTJDFNHN-UHFFFAOYSA-N | |
| 2,4,6-trimethylbenzene-1,3-diamine | |
| 76547 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 3102-70-3 | |
| 150.23 | |
| ZVDSMYGTJDFNHN-UHFFFAOYSA-N | |
| 76547 | |
| CC1=CC(C)=C(N)C(C)=C1N |
| C9H14N2 | |
| MFCD00010149 | |
| 2,4-Diaminomesitylene, 2,4-Diamino-1,3,5-trimethylbenzene | |
| 2,4,6-trimethylbenzene-1,3-diamine |
Safety and Handling
EINECSNumber : (3)-3812
RTECSNumber : CZ1607000
TSCA : No